Transfer hose out of stainless steel with PTFE piping and phase separator 3/4ft, length: 2 m 1 * 2 m
Catalog # KGWA2612-S3
Supplier: KGW
New Product
Transfer hose out of stainless steel with PTFE piping and phase separator 3/4ft, length: 2 m 1 * 2 m
Catalog # KGWA2612-S3
Supplier: KGW
New Product
Frequently Bought Together
Accessories for Dispensette®
Supplier: Brand
Accessories for HandyStep® electronic
Supplier: Brand
MOTOR 5HP 55C 115/220/50/60 1 * 1 items
Supplier: Thermo Scientific
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
SPAREPART KIT VENTIL 18 KALREZ 1 * 1 items
Supplier: KNF
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Compression fittings
Supplier: HAMILTON BONADUZ
Accessories for Diaphragm Pumps
Supplier: KNF
VAKUUM REGULATION N86/N816 1 * 1 items
Supplier: KNF
VALVE FOR AERATION LID 1 * 1 items
Supplier: Retsch GmbH
REPL COVER for CELL LOCKER COMPL. 1 * 1 items
Supplier: Thermo Scientific
LUFTBALLONE 28CM BUNT GEMISCHT 12-100er Pac
Supplier: MARKET SOURCE PART PROCESS
Fan 230V w. Blade 1 * 1 items
Supplier: Binder