Sodium Perfluoro-1-nonanesulfonate 50mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0801651.5
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Sodium Perfluoro-1-nonanesulfonate 50mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0801651.5
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test strips, Glucose, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
ACCUSPIN™ Centrifuge Tubes, Sterile, Polypropylene
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond