ContactUsLinkComponent
MSE PRO LARGE CAPACITY ROLLER JAR MILL WITH SAFETY GUARD AND DOOR INTERLOCK SYSTEM ONE TIER, 4-JAR UP TO 60L CAPACITY 1 * 1 ST
Catalog # MSPPMA0132
MSE PRO LARGE CAPACITY ROLLER JAR MILL WITH SAFETY GUARD AND DOOR INTERLOCK SYSTEM ONE TIER, 4-JAR UP TO 60L CAPACITY 1 * 1 ST
Catalog # MSPPMA0132
Frequently Bought Together