2-METHYL-5-NITROBENZALDEHYDE 1 * 5 g
Catalog # AMBDA202379-5G
Supplier: AMBEED
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl sulphoxide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Bromine ≥99.8%, liquid
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific