Order Entry
Switzerland
Orders LinkContactUsLinkComponent
High Stackable Ledges, 100 mm

High Stackable Ledges, 100 mm

Shelf ledge, Metroseal grey with Microban®, 1219 mm width
Catalog # METOL48N-4K4
Supplier:   Metro International
High Stackable Ledges, 100 mm
Catalog # METOL48N-4K4
High Stackable Ledges, 100 mm

Shelf ledge, Metroseal grey with Microban®, 1219 mm width

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Actual ledge length is approximately 25 mm shorter than nominal shelf length/width.

Specifications
  • Description:
    Shelf ledge, Metroseal grey with Microban®, 1219 mm width
  • Seq:
    07