ContactUsLinkComponent
Avantor mini-Magnetic Mixing System, 10l max capacity, heated jacket, loadcell weighing range 0-20kg 1 * 1 items
Catalog # 734-3665
Supplier:  JM SEPARATIONS
New Product
Avantor mini-Magnetic Mixing System, 10l max capacity, heated jacket, loadcell weighing range 0-20kg 1 * 1 items
Catalog # 734-3665
Supplier:  JM SEPARATIONS