Chromatography data station, DataApex ClarityVA
Clarity VA Chromatography Data Station full version. MS Data processing and DAD
Catalog # HPLCDAAPC50-18B5AC
Supplier: COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
Catalog # HPLCDAAPC50-18B5AC
Supplier: COLLECTION PREFIX HPLC
Jump to:
Product Details & Documents
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
- Direct control of Hitachi HPLC (Chromaster and Primaide)
- Modules for specific calculation and methodology
- Tools for regulated environment
- Extended user support
- Language localisations (English, German, French,...)
ClarityVA is useable for modern complained pharma labs, when configured correctly (21 CFR part 11 compliance).
Documents
Clarity Advanced Chromatography Software
Product Literature
The DataApex story
Videos
Specifications
- Description:Clarity VA Chromatography Data Station full version. MS Data processing and DAD
- Seq:03