ContactUsLinkComponent
,,26 components: 100ug/ml each of Ag Al As B Ba Bi Ca Cd Co Cr Cu Fe Ga K Li Mg Mn Na Ni Pb Sb Sn Sr Ti V Zn in HNO3 8% HF 1% ,,,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months.,, Ambient Te 1 * 100 mL
Katalognr. CMPLRM111549L1
New Product
,,26 components: 100ug/ml each of Ag Al As B Ba Bi Ca Cd Co Cr Cu Fe Ga K Li Mg Mn Na Ni Pb Sb Sn Sr Ti V Zn in HNO3 8% HF 1% ,,,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months.,, Ambient Te 1 * 100 mL
Katalognr. CMPLRM111549L1