CRYOELECTRIC COOLING SYSTEM (LN2 FREE) MTBF 100 000 HEURES NO DEGRADATION OF RESOLUTION LOW POWER CONSUMPTION 100 W CONTAINS NO FLAM MABLE GASES INCLUDES COLDHEAD (CP5CH) WITH CRYOPULSE ALIMENTATION AND CONTROLLERR (CP5CO) CP5CH DIMENSION (WITHOUT DE 1 * 1 ST
Catalog # MSPPCP5-PLUS
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together