ContactUsLinkComponent
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 200 mL
N° de catalogue ACSDRM107895L2
Fournisseur:  CPACHEM
New Product
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 200 mL
N° de catalogue ACSDRM107895L2
Fournisseur:  CPACHEM