SmarAct CLS-3232-S-C60-P34 Linear positioner, exchangeable stick-slip drive unit (EDU), 21 mm travel, crossed-roller guide, integrat ed optical encoder (S), - cable length 60 cm - 45 DSUB-15 housing, male 1 * 1 ST
Catalog # MSPP30016877
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together