ContactUsLinkComponent
Pre-stained protein ladder. Covers the size range 10-175 kDa (Tris-glycine-SDS running buffer). Number of bands = 11. Blue stained b ands at approximately 10, 40 and 90 kDa. Designed for monitoring SDS-PAGE separation and WB transfer efficiency. Thaw 1 * 500 µl
Catalog # ANTIA318977-500
Supplier:  ANTIBODIES.COM
New Product
Pre-stained protein ladder. Covers the size range 10-175 kDa (Tris-glycine-SDS running buffer). Number of bands = 11. Blue stained b ands at approximately 10, 40 and 90 kDa. Designed for monitoring SDS-PAGE separation and WB transfer efficiency. Thaw 1 * 500 µl
Catalog # ANTIA318977-500
Supplier:  ANTIBODIES.COM