Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Probes for temperature and humidity data loggers

Probes for temperature and humidity data loggers

Food probe stainless steel food sensor NTC (IP65) with TUC plug, −50…+150 °C, T: ±0.2 °C
Catalog # TEST0615.2211
Supplier:   Testo
Probes for temperature and humidity data loggers
Catalog # TEST0615.2211
Supplier:  Testo
Probes for temperature and humidity data loggers
Probes for temperature and humidity data loggers

Food probe stainless steel food sensor NTC (IP65) with TUC plug, −50…+150 °C, T: ±0.2 °C

Price
SFr. 142.00
Log in for availability
1 * 1 ST
Product Details & Documents

Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.

Specifications
  • Range:
    −50…+150 °C
  • Description:
    Food probe stainless steel food sensor NTC (IP65) with TUC plug
  • Seq:
    08
  • Accuracy:
    T: ±0.2 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.