Buffer Exchange Service (tie-in addenda to custom conjugation service) 1 * 1 items
Catalog # AATBBE001
Supplier: AAT BIOQUEST
New Product
Buffer Exchange Service (tie-in addenda to custom conjugation service) 1 * 1 items
Catalog # AATBBE001
Supplier: AAT BIOQUEST
New Product
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Test tubes, with rim
Supplier: KARTELL
VWR®, Centrifuge Tubes, Conical Bottom, PP
Supplier: VWR Collection
Petri dishes, Corning® Gosselin™
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes, without or with Dividers
Supplier: VWR Collection
Test tubes with screw caps
Supplier: KARTELL
VWR® Square Polycarbonate Media Bottles
Supplier: VWR Collection
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection
VWR® Bottles, Square, HDPE, Bar Code Labelled
Supplier: VWR Collection
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific
Centrifuge tubes, Nalgene®
Supplier: Thermo Fisher Scientific
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Gel blotting sheets, GB005, Whatman™
Supplier: Whatman products (Cytiva)
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling