Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Phenol

Phenol

100 mL
Catalog # ACSDF330654.L1
Supplier:   CPACHEM
Phenol
Catalog # ACSDF330654.L1
Supplier:  CPACHEM
CAS Number:  108-95-2
Phenol

100 mL

CAS Number:  108-95-2
Price
121.00 SFr.
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Density:1 g/cm³
Specifications

Specifications

  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.