DPC17S-V0N6-AA2 DPC17, 316 STAINLESS STEEL, VITON SEALS, NO DISPLAY, POWER 12 - 26 VDC, INPUT SIGNAL 0 - 5 VDC OUT. SIGNAL 0 - 5 VDC , RS232, WITHOUT MB & 4AB-350-SS Air 0 - 500 STANDARD ML/MIN STANDARD METERING 1 * 1 ST
Catalog # MSPPDPCS-010425
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together