ContactUsLinkComponent
15 components: 100mg/l each of As Ba Be Bi Cd Co Cr Cs Cu Ni Pb Rb Sr V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
Catalog # CMPLRM109677L1
15 components: 100mg/l each of As Ba Be Bi Cd Co Cr Cs Cu Ni Pb Rb Sr V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
Catalog # CMPLRM109677L1
Frequently Bought Together