Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories

Accessories

E 496 injector nozzle for intubation material, 4×120 mm, PG 8536
Catalog # MIEL5235490
Supplier:   MIELE
Accessories
Catalog # MIEL5235490
Supplier:  MIELE
Accessories
Accessories

E 496 injector nozzle for intubation material, 4×120 mm, PG 8536

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    E 496 injector nozzle for intubation material, 4×120 mm
  • Seq:
    14
  • For:
    PG 8536
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.