ContactUsLinkComponent
LYOPROTECT MULTIPLE USE CUP WITH SCREW-ON RING COVER VOLUME MAX. 120 ML, MATERIAL STAINLESS STEEL 316L WITH INSPECTION CERTIFICATE 3.1, MARKING GROUND WITH SERIAL NUMBER, HAND POLISHED SPECIFICATION DRAWING 1 * 1 ST
  MSPPLPMU-VS-B
LYOPROTECT MULTIPLE USE CUP WITH SCREW-ON RING COVER VOLUME MAX. 120 ML, MATERIAL STAINLESS STEEL 316L WITH INSPECTION CERTIFICATE 3.1, MARKING GROUND WITH SERIAL NUMBER, HAND POLISHED SPECIFICATION DRAWING 1 * 1 ST
  MSPPLPMU-VS-B
Frequently Bought Together