ContactUsLinkComponent
,,4 components: 5ppb each of Iodides (I-) Caesium (Cs) Molybdenum (Mo) Tellurium (Te) in Sodium Hydroxide 0.1M ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM108783L1
,,4 components: 5ppb each of Iodides (I-) Caesium (Cs) Molybdenum (Mo) Tellurium (Te) in Sodium Hydroxide 0.1M ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM108783L1
Frequently Bought Together