Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SARS Coronavirus Nucleocapsid Core Recombinant protein (340-390 aa) 1 * 0,1 mg
Catalog # PRSI39-120
Supplier:
ProSci Inc.
Expand image
SARS Coronavirus Nucleocapsid Core Recombinant protein (340-390 aa) 1 * 0,1 mg
Catalog # PRSI39-120
Supplier:
ProSci Inc.
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF