SARS NUCLEOCAPSID AB [5C6C] (HRP) 1 * 100 µl
Catalog # PRSI10-647
Supplier: ProSci Inc.
Frequently Bought Together
Measuring scoops
Supplier: VITLAB
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Cap strips, domed top
Supplier: Thermo Fisher Scientific
J.T.Baker®, Plates, Square Well
Supplier: Avantor
VWR®, Petri Dish, PS
Supplier: VWR Collection
Microcentrifuge tubes, 1,5 ml, in resealable bags, SuperSpin®
Supplier: Labcon
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Transfer pipettes
Supplier: RATIOLAB
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Sterilised write-on bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF