SARS-CoV-2 (COVID-19) Spike RBD Monoclonal Antibody [B003] (Biotin) 1 * 0,1 mg
Catalog # PRSI10-561
Supplier: ProSci Inc.
SARS-CoV-2 (COVID-19) Spike RBD Monoclonal Antibody [B003] (Biotin) 1 * 0,1 mg
Catalog # PRSI10-561
Supplier: ProSci Inc.
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Laboratory trays
Supplier: KARTELL
External thread cryogenic vials
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Adhesive PCR Foil Seals
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Centrifuge tubes, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Petri dish racks, stainless steel, PetriPile®
Supplier: INTERSCIENCE
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Storage Boxes, PP, with Lid
Supplier: KOEHLER TECHNISCHE PRODUKTEN