FAN UNIT CONDENSING UNIT 1 * 1 items
Catalog # ZIEG930412035
Anbieter: ZIEGRA EISMASCHINEN
Frequently Bought Together
SPARE PART SET SX950 1 * 1 items
Anbieter: KNF
THERMOCOUPLE TYPE K 140 MM DIAM 3 MM 1 * 1 items
Anbieter: NABERTHERM
BIOTECTOR BASE REAGENT 1 * 20 L
Anbieter: Hach
PUMP HEAD - FLEXIPUMP 1 * 1 items
Anbieter: INTERSCIENCE
BOTTLE SPRAY 250ML PE W/NOZZLE ECOLAB 1 * 1 items
Anbieter: KOEHLER TECHNISCHE PRODUKTEN
FUSE HOLDER ASSY FOR HS MODELS 1 * 1 items
Anbieter: witeg Labortechnik
THERMOSTAT 50-300°C CLASS 3.1 1 * 1 items
Anbieter: Binder
High-performance cyclone 1 * 1 items
Anbieter: BUCHI
Accessories for Rotavapor® R-3 / R-II / R-210 / R-215
Anbieter: BUCHI
6 intake manifold with CN 29/32 valve 1 * 1 items
Anbieter: Azbil Telstar Technologies
[EN]GASKET FLEX GRAPHITE 1 * 6 ST
Anbieter: PARR
Door contact switch (1S) 250VAC / 25W 1 * 1 items
Anbieter: Binder
Accessories for multi channel pipettes, mechanical, Research® plus
Anbieter: EPPENDORF
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Anbieter: Hach