Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Α-FETOPROTEIN RECOMBINANT MONOCLONAL ANTIBODY (AF5) (MOUSE IGG1Κ) 1 * 200 µG
Catalog # ENZOENZABS6070200
Supplier:
ENZO LIFE SCIENCES
Expand image
Α-FETOPROTEIN RECOMBINANT MONOCLONAL ANTIBODY (AF5) (MOUSE IGG1Κ) 1 * 200 µG
Catalog # ENZOENZABS6070200
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Pasteur pipettes, disposable
Supplier: Brand
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Petri dishes
Supplier: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF