AMPIGENE® Taq mix
AMPIGENE® Taq mix, 1 KIT
ENZOENZNUC1000040
Optimised standard PCR mix for enhanced speed, yield, and specificity. Used for standard PCR applications.
AMPIGENE® Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq Mix is a robust mix for routine PCR applications including genotyping, screening, and library construction. An optimized buffer system allows efficient amplification under fast and standard cycling conditions.
: Kit contains 2×AMPIGENE® Taq Mix (AMPIGENE® Taq Polymerase, 6 mM MgCl₂, 2 mM dNTPs, enhancer and stabilizers).
- Pk:1 KIT
- Description:AMPIGENE® Taq mix
- Cat. No.:ENZOENZNUC1000040
- Storage conditions:−20 °C