Methotrexate
Glass bottle for solids, 100 mg
ENZOALX440045M100
: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- :59-05-2
- :C₂₀H₂₂N₈O₅
- :MFCD00150847
- :454.44 g/mol
- :Freezer
- :200-413-8
- :2811
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:100 mg