Methotrexate
Glass bottle for solids, 1 g
Catalog # ENZOALX440045G001
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS Number:59-05-2
- Formula:C₂₀H₂₂N₈O₅
- MDL Number:MFCD00150847
- Molecular Weight:454.44 g/mol
- Storage Temperature:Freezer
- EINECS:200-413-8
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



