5-(4,4,5,5-TETRAMETHYL-[1,3,2]DIOXABOROLAN-2-YL)-OXAZOLE-2-CARBOXYLIC ACID ETHYL ESTER 1 * 250 mg
Catalog # JWPH32R0341P-250MG
Supplier: J W PHARMLAB
5-(4,4,5,5-TETRAMETHYL-[1,3,2]DIOXABOROLAN-2-YL)-OXAZOLE-2-CARBOXYLIC ACID ETHYL ESTER 1 * 250 mg
Catalog # JWPH32R0341P-250MG
Supplier: J W PHARMLAB
Frequently Bought Together
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Nitrobenzene 99+% ACS
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific