ContactUsLinkComponent
,,23 components: Al 500ppm Cr 500ppm Fe 500ppm Y 500ppm Zn 500ppm Ag 100ppm B 100ppm Ba 100ppm Bi 100ppm Ca 100ppm Cd 100ppm Co 100ppm Cu 100ppm K 100ppm Li 100ppm Mg 100ppm Mn 100ppm Mo 100ppm Na 100ppm Ni 100pp 1 * 100 mL
Catalog # CMPLRM009340L1
,,23 components: Al 500ppm Cr 500ppm Fe 500ppm Y 500ppm Zn 500ppm Ag 100ppm B 100ppm Ba 100ppm Bi 100ppm Ca 100ppm Cd 100ppm Co 100ppm Cu 100ppm K 100ppm Li 100ppm Mg 100ppm Mn 100ppm Mo 100ppm Na 100ppm Ni 100pp 1 * 100 mL
Catalog # CMPLRM009340L1
Frequently Bought Together