Volumetric flasks, class A, blue markings
350 ml, 14,5/23
Catalog # NEUB0293-04-035014
Supplier: NEUBERT VOLUME GLASSWAERE
Volumetric flasks, class A, blue markings
Catalog # NEUB0293-04-035014
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass, grade 3.3, with PE stopper.
- With one mark
- Good chemical resistance
- Good temperature resistance
Specifications
- NS:14,5/23
- Capacity:350 ml
- Seq:08