SCHWEFELSÄURE 37,5 % 1 * 25 kg
Catalog # MSPP1592.12
Anbieter: MARKET SOURCE PART PROCESS
Frequently Bought Together
VEPOCHEMIE Entkalker-Schaum Vapo 360 500 ml 1 * 500 mL
Anbieter: MARKET SOURCE PART PROCESS
HiPur,Acetone,4x1GAL 1 * 4 items
Anbieter: Epredia
Clearant,Xylene,4x5L 1 * 4 items
Anbieter: Epredia
[EN]IGG1 ISOTYPE CONTROL P331S/E430G 1 * 100 mg
Anbieter: MARKET SOURCE PART PROCESS
Zincon sodium salt ≥75% (dye content), Sigma-Aldrich®
Anbieter: SIGMA ALDRICH MICROSCOPY
Potassium hydroxide ≥85%, flakes
Anbieter: Thermo Fisher Scientific