Orders LinkContactUsLinkComponent
Pressure Transducers
Pressure Transducers
Pressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated, −10 - +75 psi
Catalog # SPECACPM-05TC-C1S
Pressure Transducers
Catalog # SPECACPM-05TC-C1S
Pressure Transducers
Pressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated, −10 - +75 psi
Price
Inquire For Price
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Range:
    −10 - +75 psi
  • Description:
    Pressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated
  • Seq:
    01
  • Cat. No.:
    SPECACPM-05TC-C1S
  • Accuracy:
    ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.