ContactUsLinkComponent
1 component: Phenacetin [CAS:62-44-2] 250mg/l in Ethanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 5 Ampoul
Catalog # CMPLRD1040071.5
1 component: Phenacetin [CAS:62-44-2] 250mg/l in Ethanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 5 Ampoul
Catalog # CMPLRD1040071.5
Frequently Bought Together