Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methotrexate

Methotrexate

Bottle, 10 g
Catalog # APOSBIM0130-10G
Toxic
Health
Supplier:   Apollo Scientific
Methotrexate
Catalog # APOSBIM0130-10G
Supplier:  Apollo Scientific
CAS Number:  59-05-2
Methotrexate

Bottle, 10 g

CAS Number:  59-05-2
Price
SFr. 88.40
Log in for availability
1 * 10 g
Product Details & Documents

An antineoplastic agent which acts as an antimetaboliteof folic acid.

Synonyms:  (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
  • CAS Number:59-05-2
  • Formula:C₂₀H₂₂N₈O₅
  • MDL Number:MFCD00150847
  • Molecular Weight:454.44 g/mol
  • Melting Pt:185…204 °C
  • Storage Temperature:Freezer
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications