Methotrexate
Bottle, 5 g
APOSBIM0130-5G
An antineoplastic agent which acts as an antimetaboliteof folic acid.
: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- :59-05-2
- :C₂₀H₂₂N₈O₅
- :MFCD00150847
- :454.44 g/mol
- :185…204 °C
- :Freezer
- :1544
- :6.1,III
- Pack type:Bottle
- Pk:5 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



