ContactUsLinkComponent
Chemiluminescence and Fluorescence Training Kit Includes a 12-230kDa Fluorescence Separation Module (SM FL004), Anti-Rabbit Secondar y IR Antibody, Milk-Free Antibody Diluent, Streptavidin-NIR, Anti-Mouse Secondary NIR Antibody. 1 * 1 ST
Catalog # PRTSPS-T005
Supplier:  PROTEIN SIMPLE
Chemiluminescence and Fluorescence Training Kit Includes a 12-230kDa Fluorescence Separation Module (SM FL004), Anti-Rabbit Secondar y IR Antibody, Milk-Free Antibody Diluent, Streptavidin-NIR, Anti-Mouse Secondary NIR Antibody. 1 * 1 ST
Catalog # PRTSPS-T005
Supplier:  PROTEIN SIMPLE
Frequently Bought Together