Orders LinkContactUsLinkComponent
Essential Chromatography Pump Systems, C-900 PURE
Essential Chromatography Pump Systems, C-900 PURE
C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, fraction collector (closed fraction collector bay), C-900
Catalog # 11C90020.
Supplier:   BUCHI
Essential Chromatography Pump Systems, C-900 PURE
Catalog # 11C90020.
Supplier:  BUCHI
Essential Chromatography Pump Systems, C-900 PURE
Essential Chromatography Pump Systems, C-900 PURE
C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, fraction collector (closed fraction collector bay), C-900
Price
SFr. 13’100.00
Log in for availability
1 * 1 ST
Product Details & Documents
C-900 PURE Essential chromatography pump system with flexible software control.
  • Easy accessibility and large touchscreen user interface
  • Basic design and setup
  • Intuitive handling
  • Small footprint and robust
  • Automated capabilities
  • Expansion possibilities
  • No limits for sample loading and fraction collection
The Pure Essential flash chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
Designed for simplicity, power, and safety, all within a small footprint, the Pure Essential chromatography system is ideal for crowded labs and users taking their first steps into automated chromatography. The instrument’s focus is on getting the core components right and includes a powerful 50 bar pump, an easily configurable UV detector, and a fraction collector that can be added if required.
Supporting you in every step of your research journey, the modular and scalable Pure Essential chromatography system was designed to grow with your research needs. Offering unmatched flexibility and scalability for your separation process.
 : The scope of delivery depends on the configuration of the purchase order. The pump module includes 1 Pure Chromatography C-900 (pump tower), 1 power cord, 1 solvent line pressure side kit, 2 fitting kits (one-piece fitting ⅛") and 1 operational manual.
Documents
Specifications
  • Model:
    C-900
  • Description:
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, fraction collector (closed fraction collector bay)
  • Seq:
    02