NATRIUMAZID 10 % - 500 ML ZUM KONSERVIEREN VON LABOREAGENZIEN 1 * 500 mL
MSPP13553.00500
NATRIUMAZID 10 % - 500 ML ZUM KONSERVIEREN VON LABOREAGENZIEN 1 * 500 mL
MSPP13553.00500
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals