ContactUsLinkComponent
Small parts cleaner stainless steel with recessed grip, lid closes automatically in the event of fire due to integrated fusible link release, grounding connection ideal for cleaning or degreasing small parts capacity: 8.00 litres ø x H (mm): 255 x 26 1 * 1 items
Catalog # MSGA40449
Supplier:  asecos
Small parts cleaner stainless steel with recessed grip, lid closes automatically in the event of fire due to integrated fusible link release, grounding connection ideal for cleaning or degreasing small parts capacity: 8.00 litres ø x H (mm): 255 x 26 1 * 1 items
Catalog # MSGA40449
Supplier:  asecos
Frequently Bought Together