1 COMPONENT POTASSIUM 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM021711L1
Supplier: CUSTOM MADE CHEMICALS LAB
1 COMPONENT POTASSIUM 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM021711L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
pH indicator strips, Whatman™
Supplier: Whatman products (Cytiva)
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 28%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips, glucose, Reflectoquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, peracetic acid, Reflectoquant®, Supelco®
Supplier: Merck
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Accessories
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Saffron, Q Path®
Supplier: VWR Chemicals
Starch indicator solution, MDB
Supplier: Hach
Powder pillows, sulphate, Sulfa 4
Supplier: Lovibond