Magnetic stirring bar, PTFE, triangular
12 mm, 6 mm
Catalog # COWISAN001.412
Supplier: COWIE
Jump to:
Product Details & Documents
Magnetic stirring bar which is ideal for dissolving solids and mixing sediments due to the scraper-like action on the base of the container.
Documents
Certificates
Search by Lot NumberSpecifications
- Ø:6 mm
- Length:12 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



