Injection spikes for multidose vials, Mini Spike Plus®
Injection spikes with chemo valve
BRAU4550587
: B. Braun
Mini Spike Plus withdrawal and injection spikes for multidose vials and multiple withdrawals.
- Bacteria-proof venting and comfortable handling
- Higher safety, increased efficiency, high flow rate
- Effortless and precise penetration even of thick stoppers
Integrated snap cap suitable for one-handed opening and closing with a reliable protection of drugs against microbial contamination during withdrawal.
Documents
- Description:Injection spikes with chemo valve
- Seq:05
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.