ContactUsLinkComponent
Glass Reaction Vessel, Double Walled with Inlet/Outlet Port for Heating and Cooling for Use with 2L BIOne Single Use Systems 1 * 1 items
Catalog # DISI3010-0288
Supplier:  DISTEK
Glass Reaction Vessel, Double Walled with Inlet/Outlet Port for Heating and Cooling for Use with 2L BIOne Single Use Systems 1 * 1 items
Catalog # DISI3010-0288
Supplier:  DISTEK
Frequently Bought Together