Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Azotometer/saccharometer

Azotometer/saccharometer, measuring tube alone, 50 ml, 0 − 100 ml, 1/5
Catalog # NEUB0345-51-050
Azotometer/saccharometer
Catalog # NEUB0345-51-050
Azotometer/saccharometer

Azotometer/saccharometer, measuring tube alone, 50 ml, 0 − 100 ml, 1/5

Price
SFr. 315.00
Log in for availability
1 * 1 ST
Product Details & Documents

Azotometer/saccharometer made of borosilicate glass, with glass base.

Specifications
  • Description:
    Azotometer/saccharometer, measuring tube alone, 50 ml
  • Seq:
    04
  • Division:
    1/5
  • Range:
    0 − 100 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.