ContactUsLinkComponent
,,9 components: Au 300mg/l Pd 300mg/l Ir 300mg/l Os 300mg/l Rh 300mg/l Ru 300mg/l Pt 300mg/l Sb 3600mg/l Mo 9000mg/l in HCl 10% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM104042L1
,,9 components: Au 300mg/l Pd 300mg/l Ir 300mg/l Os 300mg/l Rh 300mg/l Ru 300mg/l Pt 300mg/l Sb 3600mg/l Mo 9000mg/l in HCl 10% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM104042L1
Frequently Bought Together