,,12 components: Pb 15mg/l As 45mg/l Hg 90mg/l Co 150mg/l V 300mg/l Ni 600mg/l Tl 24mg/l Se 450mg/l Li 1650mg/l Ba 4200mg/l Cu 9000mg/l Ag 450mg/l in HNO3 5% HF TR ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, , 1 * 100 mL
CMPLRM104041L1
Frequently Bought Together