BEHROTEST SOXHLET EXTRACTION UNIT 1 * 1 items
Catalog # BEHRB00218442
Supplier: BEHR
Frequently Bought Together
VWR®, Combs for PerfectBlue™ Gel System Maxi M
Supplier: VWR Collection
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
Shut-off valves, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Two-piece fingertight fittings for ¹/₁₆" Ø tubing, SealTight™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
CRS SPure™ HC Filters
Supplier: Trajan Scientific and Medical
PEEK tubing
Supplier: Trajan Scientific and Medical
Accessories for DOSI pumps
Supplier: BioTool
Gas detection tubes for sulphur dioxide
Supplier: Dräger
PEEK TUBE NATURAL 1/8 OD X 0.062 ID, LENGTH 150 METERS 1 * 1 items
Supplier: Trajan Scientific and Medical
VWR®, Combs for PerfectBlue Gel System Mini L
Supplier: VWR Collection
Storage phosphor screens and cassettes
Supplier: Cytiva
Low pressure unions, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Environmental test chambers, MKF series
Supplier: Binder
Density meters, Densito and DensitoPro
Supplier: Mettler - Toledo