STANDARD SILICA 100PPB F/SILICA ANALYZER 1 * 1 items
ORIO223010
Frequently Bought Together
Buffer solutions in sachets
: Mettler - Toledo
pH buffers, ready to use
: WTW
ASTM Method D3710 Quantitative Calibration Standard 1 * 1 mL
: EHRENSTORFER
Colour standards
: Hach
VWR®, Flow Cup viscosity standards
: VWR Chemicals