Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
10mg/l each of Ag As Ba Cd Co Cr Cu Hg Li Ni Pb Sb Se Sn Tl V Mo 1 * 100 mL
Catalog # CMPLRM103521L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
10mg/l each of Ag As Ba Cd Co Cr Cu Hg Li Ni Pb Sb Se Sn Tl V Mo 1 * 100 mL
Catalog # CMPLRM103521L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach